missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorodiphenyltrichloroethane-13C12, TRC

Product Code. 30134196
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134196 10 mg 10mg
30005962 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134196 Supplier Toronto Research Chemicals Supplier No. TRCD43419810MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104215-84-1
Molecular Formula 13C12 C2 H9 Cl5
Molecular Weight (g/mol) 366.4
Synonym Benzene-13C6, 1,1'-(2,2,2-trichloroethylidene)bis[4-chloro- (9CI), 4,4'-DDT 13C12, 4,4'-DDT (ring 13C12): 4,4'-DDT 13C12 (benzen 13C12)
IUPAC Name 1-chloro-4-[2,2,2-trichloro-1-(4-chloro(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl)ethyl](1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene
SMILES Cl[13c]1[13cH][13cH][13c]([13cH][13cH]1)C([13c]2[13cH][13cH][13c](Cl)[13cH][13cH]2)C(Cl)(Cl)Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H9Cl5/c15-11-5-1-9(2-6-11)13(14(17,18)19)10-3-7-12(16)8-4-10/h1-8,13H/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1
Formula Weight 363.9549
Chemical Name or Material 4,4'-DDT 13C12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.