missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorodiphenyldichloroethane, TRC

Product Code. 30382381
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382381 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382381 Supplier Toronto Research Chemicals Supplier No. TRCD434185100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-DDD
CAS 72-54-8
Molecular Formula C14 H10 Cl4
Quantity 100 mg
Synonym Ethane, 1,1-dichloro-2,2-bis(p-chlorophenyl)- (8CI), 1,1'-(2,2-Dichloroethylidene)bis[4-chlorobenzene], 1,1-Bis(4-chlorophenyl)-2,2-dichloroethane, 1,1-Bis(p-chlorophenyl)-2,2-dichloroethane, 1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane, 1,1-Dichloro-2,2-bis(p-chlorophenyl)ethane, 2,2-Bis(4-chlorophenyl)-1,1-dichloroethane, 2,2-Bis(p-chlorophenyl)-1,1-dichloroethane, 2,2-Di(p-chlorophenyl)-1,1-dichloroethane, 4,4'-1,1-Dichloro-2,2-bis(p-chlorophenyl)ethane, 4,4'-DDD, 4,4'-Dichlorodiphenyldichloroethane, 4,4'-TDE, DDD, Dichlorodiphenyldichloroethane, Dilene, ME 1700, NSC 8941, Rhothane, TDE, p,p'-DDD, p,p'-Dichlorodiphenyldichloroethane, p,p'-Dichlorodiphenylethylene dichloride, p,p'-TDE
InChI Formula InChI=1S/C14H10Cl4/c15-11-5-1-9(2-6-11)13(14(17)18)10-3-7-12(16)8-4-10/h1-8,13-14H
SMILES ClC(Cl)C(c1ccc(Cl)cc1)c2ccc(Cl)cc2
IUPAC Name 1-chloro-4-[2,2-dichloro-1-(4-chlorophenyl)ethyl]benzene
Molecular Weight (g/mol) 320.04
Formula Weight 317.9537
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.