missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorodiphenyl sulfone, 97%, Thermo Scientific Chemicals

Catalog Number p-105284
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
80-07-9
C12H8Cl2O2S
287.15
MFCD00000619
GPAPPPVRLPGFEQ-UHFFFAOYSA-N
4,4'-dichlorodiphenyl sulfone, bis 4-chlorophenyl sulfone, p,p'-dichlorodiphenyl sulfone, sulfone, bis p-chlorophenyl, 4-chlorophenyl sulfone, 1,1'-sulfonylbis 4-chlorobenzene, bis p-chlorophenyl sulfone, benzene, 1,1'-sulfonylbis 4-chloro, p-chlorophenyl sulfone, 4,4'-dichlorodiphenylsulfone
6625
1-chloro-4-(4-chlorophenyl)sulfonylbenzene
ClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1
This item is not returnable. View return policy
80-07-9
C12H8Cl2O2S
287.15
MFCD00000619
GPAPPPVRLPGFEQ-UHFFFAOYSA-N
4,4'-dichlorodiphenyl sulfone, bis 4-chlorophenyl sulfone, p,p'-dichlorodiphenyl sulfone, sulfone, bis p-chlorophenyl, 4-chlorophenyl sulfone, 1,1'-sulfonylbis 4-chlorobenzene, bis p-chlorophenyl sulfone, benzene, 1,1'-sulfonylbis 4-chloro, p-chlorophenyl sulfone, 4,4'-dichlorodiphenylsulfone
6625
1-chloro-4-(4-chlorophenyl)sulfonylbenzene
ClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 146.0°C to 151.0°C
Color White
Boiling Point 397.0°C
Flash Point 233°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 25 g
Beilstein 06, 327
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohols,slightly soluble in aqueous solvents
Formula Weight 287.16
Percent Purity 97%
Physical Form Fine Powder
Chemical Name or Material 4, 4'-Dichlorodiphenyl sulfone, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'