missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorodiphenyl Sulfone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30327421
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327421 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327421 Supplier Dr. Ehrenstorfer Supplier No. DREA12421800AL100
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-Dichlorodiphenyl Sulfone
CAS 80-07-9
Analyte or Component Names 4,4'-Dichlorodiphenyl Sulfone
Shipping Condition Room Temperature
Molecular Formula C12 H8 Cl2 O2 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Sulfone, bis(p-chlorophenyl) (6CI,8CI), 1,1'-Sulfonylbis[4-chlorobenzene], 4,4'-Dichlorodiphenyl sulfone, 4-Chloro-1-(4-chlorophenylsulfonyl)benzene, 4-Chlorophenyl sulfone, B 0810, Bis(4-chlorophenyl) sulfone, Bis(p-chlorophenyl) sulfone, Di-p-chlorophenyl sulfone, NSC 23899, NSC 38759, NSC 50730, NSC 7207, p,p'-Dichlorodiphenyl sulfone, p-Chlorophenyl sulfone, Benzene, 1,1'-sulfonylbis[4-chloro-, 4,4'-Dichlorodiphenyl Sulfone
InChI Formula InChI=1S/C12H8Cl2O2S/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12/h1-8H
SMILES Clc1ccc(cc1)S(=O)(=O)c2ccc(Cl)cc2
IUPAC Name 1-chloro-4-(4-chlorophenyl)sulfonylbenzene
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 287.16
Quantity 1 mL
Formula Weight 285.9622 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.