missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorobutyrophenone, 97%, Thermo Scientific™

Product Code. 10193660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10193660 100 g 100g
10396490 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10193660 Supplier Thermo Scientific Acros Supplier No. 136891000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 40877-09-6
Molecular Formula C10H10Cl2O
Molecular Weight (g/mol) 217.09
MDL Number MFCD00018993
InChI Key UKCHLVFIVJBCKE-UHFFFAOYSA-N
Synonym 4-chloro-1-4-chlorophenyl butan-1-one, 4,4'-dichlorobutyrophenone, 1-butanone, 4-chloro-1-4-chlorophenyl, gamma,p-dichlorobutyrophenone, 4-chloro-1-4-chlorophenyl-1-oxobutane, 4-chloro-1-4-chlorophenyl-1-butanone, acmc-20a5su, 4',gamma-dichlorobutyrophenone, 4-chloro-4'-chlorobutyrophenone, 4-chloro-4'-chloro-butyrophenone
PubChem CID 2723781
IUPAC Name 4-chloro-1-(4-chlorophenyl)butan-1-one
SMILES C1=CC(=CC=C1C(=O)CCCCl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 29°C to 30°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula ClC6H4CO(CH2)3Cl
Quantity 100 g
Solubility Information Solubility in water: insoluble
Formula Weight 217.09
Percent Purity 97%
Physical Form Pellets
Chemical Name or Material 4, 4'-Dichlorobutyrophenone, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.