missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromooctafluorobiphenyl, 99%

Product Code. 11496167
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496167 1 g 1g
11406177 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496167 Supplier Thermo Scientific Alfa Aesar Supplier No. B25126.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10386-84-2 | C12Br2F8 | 455.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10386-84-2
Molecular Formula C12Br2F8
Molecular Weight (g/mol) 455.93
MDL Number MFCD00000310
InChI Key YXLMNFVUNLCJJY-UHFFFAOYSA-N
PubChem CID 82600
IUPAC Name 1-bromo-4-(4-bromo-2,3,5,6-tetrafluorophenyl)-2,3,5,6-tetrafluorobenzene
SMILES FC1=C(F)C(=C(F)C(F)=C1Br)C1=C(F)C(F)=C(Br)C(F)=C1F

Specifications

Melting Point 113°C to 115°C
Quantity 1 g
UN Number UN3152
Synonym 4,4'-dibromooctafluorobiphenyl, 4,4'-dibromo-2,2',3,3',5,5',6,6'-octafluorobiphenyl, octafluoro-4,4'-dibromobiphenyl, 4,4'-dibromo-2,2',3,3',5,5',6,6'-octafluoro-1,1'-biphenyl, 1,1'-biphenyl, 4,4'-dibromo-2,2',3,3',5,5',6,6'-octafluoro, 1,1'-biphenyl,4,4'-dibromo-2,2',3,3',5,5',6,6'-octafluoro, 1-bromo-4-4-bromo-2,3,5,6-tetrafluorophenyl-2,3,5,6-tetrafluorobenzene, f-pbb, 4,4/'-dibromooctafluorobiphenyl, 4,4\\'-dibromooctafluorobiphenyl
Formula Weight 455.94
Percent Purity 99%
Chemical Name or Material 4,4'-Dibromooctafluorobiphenyl
Product Identifier
  • 4,4'-Dibromooctafluorobiphenyl
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 4,4-Dibromo-2,2,3,3,5,5,6,6-octafluoro-1,1-biphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.