missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromobiphenyl, 98+%

Product Code. 11468106
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468106 50 g 50g
11478106 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11468106 Supplier Thermo Scientific Alfa Aesar Supplier No. A14139.18
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 92-86-4 | C12H8Br2 | 312.00 g/mol

4,4'-Dibromobiphenyl is commonly used as fire retardants and plasticizers. It is also have application in gas chromatography and liquid chromatography analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dibromobiphenyl is commonly used as fire retardants and plasticizers. It is also have application in gas chromatography and liquid chromatography analysis.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-86-4
Molecular Formula C12H8Br2
Molecular Weight (g/mol) 312.00
MDL Number MFCD00000101
InChI Key HQJQYILBCQPYBI-UHFFFAOYSA-N
Synonym 4,4'-dibromobiphenyl, 1,1'-biphenyl, 4,4'-dibromo, p,p'-dibromobiphenyl, 4,4'-dibromodiphenyl, 1-bromo-4-4-bromophenyl benzene, 4,4'-dibromo-biphenyl, unii-pou26v2xt2, 4',4-dibromobiphenyl, biphenyl, 4,4'-dibromo, pou26v2xt2
PubChem CID 7110
IUPAC Name 1-bromo-4-(4-bromophenyl)benzene
SMILES BrC1=CC=C(C=C1)C1=CC=C(Br)C=C1

Specifications

Melting Point 164°C to 169°C
Boiling Point 355°C to 360°C
Quantity 50 g
UN Number UN3152
Beilstein 2044701
Solubility Information Insoluble in water.
Formula Weight 312.02
Percent Purity ≥98%
Chemical Name or Material 4,4'-Dibromobiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.