missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromobenzil, 97%, Thermo Scientific Chemicals

£38.60 - £578.00

Chemical Identifiers

CAS 35578-47-3
Molecular Formula C14H8Br2O2
MDL Number MFCD00000104
InChI Key NYCBYBDDECLFPE-UHFFFAOYSA-N
PubChem CID 96430
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11454414
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13090.09
10 g
£38.60
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11464414
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13090.18
50 g
£152.00
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11474414
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13090.30
250 g
£578.00
250g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4,4?-Dibromobenzil was used in preparation of 5,5?-bis -(4-bromo-phenyl)-3-methyl-imidazolidine-2,4-dione, 5,5?- bis -(4-bromo-phenyl)-3-butyl-imidazolidine-2,4-dione, 5,5?-bis-(4-bromo-phenyl)-3-pentyl-imidazolidine-2,4-dione and 5,5?-bis -(4-chloro-phenyl)-3-ethyl imidazolidine-2,4-dione. It was also used in synthesis of novel quinoxaline-based sensitizers for dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dibromobenzil was used in preparation of 5,5′-bis -(4-bromo-phenyl)-3-methyl-imidazolidine-2,4-dione, 5,5′- bis -(4-bromo-phenyl)-3-butyl-imidazolidine-2,4-dione, 5,5′-bis-(4-bromo-phenyl)-3-pentyl-imidazolidine-2,4-dione and 5,5′-bis -(4-chloro-phenyl)-3-ethyl imidazolidine-2,4-dione. It was also used in synthesis of novel quinoxaline-based sensitizers for dye-sensitized solar cells.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

35578-47-3
MFCD00000104
96430
C14H8Br2O2
NYCBYBDDECLFPE-UHFFFAOYSA-N

Specifications

35578-47-3
C14H8Br2O2
4,4'-dibromobenzil, ethanedione, bis 4-bromophenyl, p,p'-dibromobenzil, 1,2-bis 4-bromophenyl ethane-1,2-dione, benzyl, 4,4'-dibromo, unii-o4txv8a656, bis 4-bromophenyl ethanedione, 1,2-ethanedione, 1,2-bis 4-bromophenyl, bis 4-bromophenyl ethane-1,2-dione, 1,2-bis-4-bromo-phenyl-ethane-1,2-dione
NYCBYBDDECLFPE-UHFFFAOYSA-N
1,2-bis(4-bromophenyl)ethane-1,2-dione
96430
97%
224°C to 226°C
MFCD00000104
Sparingly soluble in water.
C1=CC(=CC=C1C(=O)C(=O)C2=CC=C(C=C2)Br)Br
368.024
368.04
4,4'-Dibromobenzil
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 252-628-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only