missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Di-tert-butyldiphenylmethane, 99%, Thermo Scientific™

Product Code. 11494847
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11494847

Brand: Thermo Scientific Alfa Aesar B21820.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediate for pharmaceuticals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19099-48-0
Molecular Formula C21H28
Molecular Weight (g/mol) 280.455
MDL Number MFCD00043733
InChI Key KRMBVBPNWUBFBC-UHFFFAOYSA-N
Synonym 4,4'-di-tert-butyldiphenylmethane, 1-tert-butyl-4-4-tert-butylphenyl methyl benzene, bis 4-tert-butyl phenyl methane, 4,4<<−di-tert-butyldiphenylmethane, 1,1'-methylenebis 4-tert-butylbenzene, 1-tert-butyl-4-4-tert-butylbenzyl benzene, 1-tert-butyl-4-4-tert-butyl phenyl methyl benzene
PubChem CID 1652739
IUPAC Name 1-tert-butyl-4-[(4-tert-butylphenyl)methyl]benzene
SMILES CC(C)(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)C(C)(C)C

Specifications

Melting Point 71°C to 73°C
Quantity 50 g
Formula Weight 280.45
Percent Purity 99%
Chemical Name or Material 4,4'-Di-tert-butyldiphenylmethane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.