missing translation for 'onlineSavingsMsg'
Learn More

4-[(4-chlorophenyl)thio]thiophene-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. p-2701096
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10364382 250 mg 250mg
10232192 1 g 1g
10240445 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10364382 Supplier Thermo Scientific Maybridge Supplier No. KM04905CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 149609-86-9
Molecular Formula C11H7ClO2S2
Molecular Weight (g/mol) 270.745
MDL Number MFCD00052149
InChI Key UKDSGGJQRSNTHC-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl thio thiophene-3-carboxylic acid, 3-thiophenecarboxylicacid, 4-4-chlorophenyl thio, 4-4-chlorophenyl sulfanyl thiophene-3-carboxylic acid, 4-4-chlorophenyl sulfanylthiophene-3-carboxylic acid, maybridge3_003733, acmc-20n5rn, 4-4-chlorophenylsulfanyl thiophene-3-carboxylic acid, 4-4-chlorophenyl thio-3-thiophenecarboxylic acid, 4-4-chlorophenyl sulphanyl thiophene-3-carboxylic acid
PubChem CID 2778946
IUPAC Name 4-(4-chlorophenyl)sulfanylthiophene-3-carboxylic acid
SMILES C1=CC(=CC=C1SC2=CSC=C2C(=O)O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 270.76
Percent Purity 97%
Chemical Name or Material 4-[(4-Chlorophenyl)thio]thiophene-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.