missing translation for 'onlineSavingsMsg'
Learn More

4-(4-chlorophenyl)pyrimidine-2-thiol, 97%, Thermo Scientific™

Product Code. 10232552
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10232552 10 g Amber Glass Bottle 10g
10723633 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10232552 Supplier Thermo Scientific Maybridge Supplier No. KM05717EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175203-08-4
Molecular Formula C10H7ClN2S
Molecular Weight (g/mol) 222.69
MDL Number MFCD00174262
InChI Key TWKIWOOKHPWUIM-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl pyrimidine-2-thiol, 4-4-chlorophenyl-2-pyrimidinethiol, 4-4-chloro-phenyl-pyrimidine-2-thiol, 6-4-chlorophenyl-1h-pyrimidine-2-thione, 2 1h-pyrimidinethione, 4-4-chlorophenyl, maybridge1_004888, 6-4-chlorophenyl pyrimidine-2 1h-thione, 4-4-chlorophenyl-3h-pyrimidine-2-thione
PubChem CID 2735805
IUPAC Name 6-(4-chlorophenyl)-1H-pyrimidine-2-thione
SMILES ClC1=CC=C(C=C1)C1=CC=NC(=S)N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 222.7
Percent Purity 97%
Chemical Name or Material 4-(4-Chlorophenyl)pyrimidine-2-thiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.