missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Chlorophenyl)cyclohexanecarboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Linear Formula | ClC6H4C6H10CO2H |
Formula Weight | 238.71 |
Percent Purity | 97% |
Quantity | 10 g |
Packaging | Amber Glass Bottle |
Chemical Name or Material | (E)-4-(4-Chlorophenyl)cyclohexanecarboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'