missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorophenyl)-4-hydroxypiperidine, 99+%, crystalline

Product Code. 10129793 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10129793 25 g Glass bottle 25g
10309020 1 g Glass bottle 1g
10319020 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10129793 Supplier Thermo Scientific Acros Supplier No. 296840250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 39512-49-7 | C11H15Cl2NO | 248.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39512-49-7
Molecular Formula C11H15Cl2NO
Molecular Weight (g/mol) 248.15
MDL Number MFCD00006001
InChI Key PXAJMBHRLGKPQV-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl piperidin-4-ol, 4-4-chlorophenyl-4-hydroxypiperidine, 4-4-chlorophenyl-4-piperidinol, 4-chlorophenyl-4-hydroxylpiperidine, 4-p-chlorophenyl-4-hydroxypiperidine, 4-p-chlorophenyl piperidin-4-ol, 4-piperidinol, 4-4-chlorophenyl, haloperidol metabolite i, unii-und92fks0w, 4-hydroxy-4-4-chlorophenyl piperidine
PubChem CID 38282
IUPAC Name 4-(4-chlorophenyl)piperidin-4-ol
SMILES [H+].[Cl-].OC1(CCNCC1)C1=CC=C(Cl)C=C1

Specifications

Absorbance (1 w/v% soln. in DMF) (1 cm silica cell), (1 w/v% soln. in DMF) (1 cm silica cell) at 0nm, 0.05 max. at 440nm
CAS Min % 99.0
CAS Max % 100.0
Melting Point 139°C to 141°C
Color Cream-White to White
Loss on Drying 0.5% max. (1 g, 105°C, 2 hrs, 760mm Hg)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: 340mg/l (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,dmf,,acetone and dichloromethane
Formula Weight 211.69
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Chlorophenyl)-4-hydroxypiperidine
Show More Show Less
compliance-icons
Product Identifier
  • 4-(4-chlorophenyl)piperidin-4-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.