missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorobenzoyl)pyridine, 99+%

Product Code. 11465117
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465117 1 g 1g
11475117 5 g 5g
11485117 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465117 Supplier Thermo Scientific Alfa Aesar Supplier No. B22551.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 14548-48-2 | C12H8ClNO | 217.652 g/mol

4-(4-Chlorobenzoyl)pyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Chlorobenzoyl)pyridine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14548-48-2
Molecular Formula C12H8ClNO
Molecular Weight (g/mol) 217.652
MDL Number MFCD00006431
InChI Key YMTFKKLFLLAFNI-UHFFFAOYSA-N
Synonym 4-4-chlorobenzoyl pyridine, 4-chlorophenyl pyridin-4-yl methanone, 4-4'-chlorobenzoyl pyridine, 4-chlorophenyl-pyridin-4-ylmethanone, methanone, 4-chlorophenyl-4-pyridinyl, acmc-1bu0c, 4-4-chloro-benzoyl pyridine, 4-chlorophenyl 4-pyridyl ketone, 4-chlorophenyl pyridin-4-ylmethanone, 4-chlorophenyl-4-pyridyl methanone
PubChem CID 98503
IUPAC Name (4-chlorophenyl)-pyridin-4-ylmethanone
SMILES C1=CC(=CC=C1C(=O)C2=CC=NC=C2)Cl

Specifications

Melting Point 107°C to 109°C
Quantity 1 g
Formula Weight 217.66
Percent Purity ≥99%
Chemical Name or Material 4-(4-Chlorobenzoyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.