missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorobenzoyl)piperidine, 97%, Thermo Scientific™

Product Code. 15402059
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate. The derivative of this compound, 2-amino-3-(4-chlorobenzoyl)-4-[4-(alkyl/aryl)piperazin-yl]thiophene are used as allosteric enhancers of the A1-adenosine receptor.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53220-41-0
Molecular Formula C12H14ClNO
Molecular Weight (g/mol) 223.70
MDL Number MFCD04113538
InChI Key IYGWDOXHCPQXKN-UHFFFAOYSA-N
Synonym 4-4-chlorobenzoyl piperidine, 4-chlorophenyl piperidin-4-yl methanone, 4-4-chlorophenyl carbonyl piperidine, 4-chlorophenyl-4-piperidinyl methanone, 4-chlorophenyl-piperidin-4-yl-methanone, maybridge1_003174, 4-chlorophenyl 4-piperidinyl methanonehydrochloride, 4-4-chlorobenzoyl-piperidine, 4-chlorophenyl 4-piperidyl ketone, 4-chlorophenyl piperidin-4-ylmethanone
PubChem CID 2777115
SMILES ClC1=CC=C(C=C1)C(=O)C1CCNCC1

Specifications

Melting Point 60°C to 63°C
Color Yellow
Quantity 250 mg
Beilstein 158791
Solubility Information Slightly soluble in water.
IUPAC Name (4-chlorophenyl)-piperidin-4-ylmethanone
Formula Weight 223.7
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(4-Chlorobenzoyl)piperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'