missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromophenyl)imidazole, 97%

Product Code. 15404177
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15404177 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15404177 Supplier Thermo Scientific Alfa Aesar Supplier No. H31581.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13569-96-5 | C9H7BrN2 | 223.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13569-96-5
Melting Point 138°C to 142°C
Molecular Formula C9H7BrN2
MDL Number MFCD03312935
Quantity 5 g
Synonym 4-4-bromophenyl-1h-imidazole, 1h-imidazole, 5-4-bromophenyl, 5-4-bromophenyl-1h-imidazole, 4-4-bromophenyl-3h-imidazole, cyp144a1 ligand, 4, 4-4-bromophenyl imidazole, 5-4-bromophenyl-3h-imidazole, 1h-imidazole,5-4-bromophenyl, 4-4-bromo-phenyl-1h-imidazole
InChI Key YLIOAWKNPLJMID-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C2=CN=CN2)Br
IUPAC Name 5-(4-bromophenyl)-1H-imidazole
Molecular Weight (g/mol) 223.073
PubChem CID 818595
Formula Weight 223.07
Percent Purity 97%
Chemical Name or Material 4-(4-Bromophenyl)imidazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.