missing translation for 'onlineSavingsMsg'
Learn More

4'-(4-Bromophenyl)acetophenone, 97%

Product Code. 11353257
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11353257

Brand: Thermo Scientific Alfa Aesar L13798.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5731-01-1 | C14H11BrO | 275.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5731-01-1
MDL Number MFCD00143242

Specifications

Melting Point 125°C to 127°C
Molecular Formula C13H8FO2
Quantity 100 g
Beilstein 2048590
Synonym 4-4-fluorophenyl benzoic acid, 4'-fluoro-biphenyl-4-carboxylic acid, 4'-fluorobiphenyl-4-carboxylic acid, 4'-fluoro-1,1'-biphenyl-4-carboxylic acid, 4-biphenyl-4'-fluoro-carboxylic acid, 4'-fluoro-biphenyl-4-carboxylic acid, 4-fluoro-biphenyl-4-carboxylic acid, 4'-fluoro 1,1'-biphenyl-4-carboxylic acid, 4fc
InChI Key LXWNTLBMNCXRQN-UHFFFAOYSA-M
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)F)C(=O)O
IUPAC Name 4-(4-fluorophenyl)benzoic acid
Molecular Weight (g/mol) 215.20
PubChem CID 2782689
Formula Weight 275.15
Percent Purity 97%
Chemical Name or Material 4'-(4-Bromophenyl)acetophenone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.