missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromo-2-fluorobenzyl)morpholine, 96%, Thermo Scientific™

Product Code. 15489198
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15489198

Brand: Thermo Scientific Alfa Aesar H62716.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338454-98-1
Molecular Formula C11H13BrFNO
Molecular Weight (g/mol) 274.133
MDL Number MFCD12022322
InChI Key OQMLJVPTCMLEGO-UHFFFAOYSA-N
Synonym 4-4-bromo-2-fluorobenzyl morpholine, 4-4-bromo-2-fluorophenyl methyl morpholine, acmc-209i3b, 4-4-bromo-2-fluoro-benzyl-morpholine, morpholine, 4-4-bromo-2-fluorophenyl methyl
PubChem CID 21914503
IUPAC Name 4-[(4-bromo-2-fluorophenyl)methyl]morpholine
SMILES C1COCCN1CC2=C(C=C(C=C2)Br)F

Specifications

Quantity 5 g
Formula Weight 274.13
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-(4-Bromo-2-fluorobenzyl)morpholine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.