missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester, 95%

Product Code. 15439315
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15439315 1 g 1g
15449315 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15439315 Supplier Thermo Scientific Alfa Aesar Supplier No. H51743.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 936694-19-8 | C22H35BN2O4 | 402.342 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 936694-19-8
Molecular Formula C22H35BN2O4
Molecular Weight (g/mol) 402.342
MDL Number MFCD16294502
InChI Key AAEYFMAHSYKHGD-UHFFFAOYSA-N
Synonym tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperazine-1-carboxylate, tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperazine-1-carboxylate, 4-4-boc-piperazinomethyl phenylboronic acid, pinacol ester, 4-4-boc-1-piperazinylmethyl benzeneboronic acid pinacol ester, amtb122, 4-4-boc-piperazinemethyl phenylboronic acid pinacol ester, 4-4-boc-piperazine methyl phenylboronic acid pinacol ester, tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzyl piperazine-1-carboxylate
PubChem CID 53216820
IUPAC Name tert-butyl 4-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperazine-1-carboxylate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN3CCN(CC3)C(=O)OC(C)(C)C

Specifications

Quantity 1 g
Beilstein 18669672
Formula Weight 402.34
Percent Purity 95%
Chemical Name or Material 4-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.