missing translation for 'onlineSavingsMsg'
Learn More

4,4-Bis(diethylamino)benzophenone, Dr. Ehrenstorfer

Product Code. 30324264
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324264 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324264 Supplier Dr. Ehrenstorfer Supplier No. DREC10651850
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4-Bis(diethylamino)benzophenone
CAS 90-93-7
Analyte or Component Names 4,4-Bis(diethylamino)benzophenone
Shipping Condition Room Temperature
Molecular Formula C21 H28 N2 O
Certifications/Compliance ISO/IEC 17025
Synonym Benzophenone, 4,4'-bis(diethylamino)- (6CI,7CI,8CI), Bis[4-(diethylamino)phenyl]methanone, 4,4-Bis(diethylamino)benzophenone, 4,4'-(Tetraethyldiamino)benzophenone, 4,4'-Bis(N,N-diethylamino)benzophenone, 4,4'-Bis(diethylamine)benzophenone, 4,4'-Bis(diethylamino)benzophenone, 4,4'-Bis(diethylamino)diphenyl ketone, 4,4'-Diethylaminobenzophenone, 4',4-Bis(diethylamino)benzophenone, Doublecure EMK, EAB, EAB (benzophenone), EAB-SS, EABF, Ethyl Michler ketone, JRCure EMK, Kayacure EAB, Michler's ethyl ketone, N,N,N',N'-Tetraethyl-4,4'-diaminobenzophenone, NSC 36365, Narucure CS, SB-PI 701, p,p'-Bis(diethylamino)benzophenone, p,p'-Tetraethyldiaminobenzophenone
InChI Formula InChI=1S/C21H28N2O/c1-5-22(6-2)19-13-9-17(10-14-19)21(24)18-11-15-20(16-12-18)23(7-3)8-4/h9-16H,5-8H2,1-4H3
SMILES CCN(CC)c1ccc(cc1)C(=O)c2ccc(cc2)N(CC)CC
IUPAC Name bis[4-(diethylamino)phenyl]methanone
Molecular Weight (g/mol) 324.46
Quantity 250 mg
Formula Weight 324.2202 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.