missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(3-aminophenoxy)diphenyl sulfone, 95%, Thermo Scientific™

Product Code. 11473407
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473407 1000 g 1000g
11493407 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473407 Supplier Thermo Scientific Alfa Aesar Supplier No. A17194.0B

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30203-11-3
Molecular Formula C24H20N2O4S
Molecular Weight (g/mol) 432.49
MDL Number MFCD00054738
InChI Key WCXGOVYROJJXHA-UHFFFAOYSA-N
Synonym 3,3'-sulfonylbis 4,1-phenylene bis oxy dianiline, bis 4-3-aminophenoxy phenyl sulfone, 4,4'-bis 3-aminophenoxy diphenyl sulfone, benzenamine, 3,3'-sulfonylbis 4,1-phenyleneoxy bis, 4,4-bis 3-aminophenoxy diphenyl sulfone, 3-4-4-3-aminophenoxy benzenesulfonyl phenoxy aniline, 4-3-aminophenoxy-1-4-3-aminophenoxy phenyl sulfonyl benzene, acmc-209hds, 3,3'-sulfonylbis 4,1-phenyleneoxy dianiline
PubChem CID 121657
IUPAC Name 3-[4-[4-(3-aminophenoxy)phenyl]sulfonylphenoxy]aniline
SMILES NC1=CC(OC2=CC=C(C=C2)S(=O)(=O)C2=CC=C(OC3=CC(N)=CC=C3)C=C2)=CC=C1

Specifications

Melting Point 132°C to 135°C
Quantity 1000 g
Formula Weight 432.5
Percent Purity 95%
Chemical Name or Material 4,4'-Bis(3-aminophenoxy)diphenyl sulfone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.