missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bipyridine, 98%

Product Code. 11469492
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469492 100 g 100g
11449492 5 g 5g
11459492 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469492 Supplier Thermo Scientific Alfa Aesar Supplier No. A10187.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 553-26-4 | C10H8N2 | 156.19 g/mol

4,4'-Bipyridine is used in transition-metal complex catalyst chemistry for uniform polymerization, in luminescence chemistry and in spectrophotometric analysis. It plays an important role as a photosensitizer and luminescent material. It is also used as a precursor to paraquat viz. N,N'-dimethyl-4,4'-bipyridinium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Bipyridine is used in transition-metal complex catalyst chemistry for uniform polymerization, in luminescence chemistry and in spectrophotometric analysis. It plays an important role as a photosensitizer and luminescent material. It is also used as a precursor to paraquat viz. N,N′-dimethyl-4,4′-bipyridinium.

Solubility
Soluble in ethanol, ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-26-4
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00006416
InChI Key MWVTWFVJZLCBMC-UHFFFAOYSA-N
Synonym 4,4'-bipyridine, 4,4'-bipyridyl, 4,4'-dipyridyl, 4-4-pyridyl pyridine, 4,4'-dipyridine, 4,4-bipyridyl, 4,4-dipyridyl, gamma,gamma'-bipyridyl, gamma,gamma'-dipyridyl, unii-x4o2od61cb
PubChem CID 11107
ChEBI CHEBI:30985
IUPAC Name 4-pyridin-4-ylpyridine
SMILES C1=CC(=CC=N1)C1=CC=NC=C1

Specifications

Melting Point 111°C to 114°C
Boiling Point 304°C to 305°C
Quantity 100 g
UN Number UN2811
Beilstein 113176
Sensitivity Hygroscopic
Merck Index 14,3348
Solubility Information Soluble in ethanol,ether. Insoluble in water.
Formula Weight 156.19
Percent Purity 98%
Chemical Name or Material 4,4'-Bipyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4,4'-Bipyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: 4,4-Bipyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.