missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bipiperidine dihydrochloride, 99%

Product Code. 11302747
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11302747

Brand: Thermo Scientific Alfa Aesar L15696.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 78619-84-8 | C10H22Cl2N2 | 241.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78619-84-8
Molecular Formula C10H22Cl2N2
Molecular Weight (g/mol) 241.2
MDL Number MFCD00005998
InChI Key MQKBVVIPCLTENX-UHFFFAOYSA-N
Synonym 4,4'-bipiperidine dihydrochloride, 4,4'-bipiperidyl dihydrochloride, 4,4'-bipiperidine di hydrochloride, pubchem7721, 4,4-bipiperidine dihydrochloride, 4-piperidin-4-yl piperidine dihydrochloride, 4,4 inverted exclamation marka-bipiperidine dihydrochloride, 4,4 inverted exclamation marka-bipiperidyl dihydrochloride
PubChem CID 12270637
IUPAC Name 4-piperidin-4-ylpiperidine;dihydrochloride
SMILES C1CNCCC1C2CCNCC2.Cl.Cl

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 3687878
Sensitivity Hygroscopic
Formula Weight 241.21
Percent Purity 99%
Chemical Name or Material 4,4'-Bipiperidine dihydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.