missing translation for 'onlineSavingsMsg'
Learn More

4-(4'-Aminobenzyl)thiomorpholine 1,1-dioxide, ≥90%, Thermo Scientific™

Product Code. 10292932
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10292932 1 g 1g
10384482 250 mg 250mg
10108504 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10292932 Supplier Thermo Scientific Maybridge Supplier No. MO00124DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Identificadores de productos químicos

CAS 263339-24-8
Molecular Formula C11H16N2O2S
Molecular Weight (g/mol) 240.32
MDL Number MFCD03407300
InChI Key TZMCILRCGMFRJC-UHFFFAOYSA-N
Synonym 4-4'-aminobenzyl thiomorpholine 1,1-dioxide, 4-1,1-dioxo-1,4-thiazinan-4-yl methyl aniline, 4-1,1-dioxo-4-thiomorpholinyl methyl phenyl amine, 4-4-aminophenyl methyl-1??-thiomorpholine-1,1-dione, 4-aminobenzyl thiomorpholine 1,1-dioxide, 4-4-aminobenzyl thiomorpholine 1,1-dioxide, 4-4-aminobenzyl-thiomorpholine-1,1-dioxide, 4-4-amino-benzyl-thiomorpholine 1,1-dioxide, 4-1,1-dioxo-thiomorpholin-4-yl-methyl-aniline
PubChem CID 2779682
IUPAC Name 4-[(1,1-dioxo-1,4-thiazinan-4-yl)methyl]aniline
SMILES NC1=CC=C(CN2CCS(=O)(=O)CC2)C=C1

Especificaciones

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 240.33
Percent Purity 90%
Chemical Name or Material 4-(4'-Aminobenzyl)thiomorpholine 1, 1-dioxide
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.