missing translation for 'onlineSavingsMsg'
Learn More

4-(4,6-Dimethoxy-s-triazin-2-yl)-4-methyl-morpholinium Chloride, TRC

Product Code. 30373316
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30373316 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 30373316

Brand: Toronto Research Chemicals TRCD46124550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(4,6-Dimethoxy-s-triazin-2-yl)-4-methyl-morpholinium Chloride
CAS 3945-69-5
Molecular Formula C10 H17 N4 O3 . Cl
Quantity 50 g
Synonym 4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium Chloride, DMTMM
InChI Formula InChI=1S/C10H17N4O3.ClH/c1-14(4-6-17-7-5-14)8-11-9(15-2)13-10(12-8)16-3;/h4-7H2,1-3H3;1H/q+1;/p-1
SMILES [Cl-].COc1nc(OC)nc(n1)[N+]2(C)CCOCC2
IUPAC Name 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholin-4-ium;chloride
Molecular Weight (g/mol) 276.72
Formula Weight 276.0989
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.