missing translation for 'onlineSavingsMsg'
Learn More

4-{[4,6-Bis(octylsulfanyl)-1,3,5-triazin-2-yl]amino-2,6-di-tert-butylphenol, TRC

Product Code. 30132572
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132572 500 mg 500mg
30005950 100 mg 100mg
30005270 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132572 Supplier Toronto Research Chemicals Supplier No. TRCB591828500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 991-84-4
Molecular Formula C33 H56 N4 O S2
Molecular Weight (g/mol) 588.95
Synonym Phenol, 4-[[4,6-bis(octylthio)-1,3,5-triazin-2-yl]amino]-2,6-bis(1,1-dimethylethyl)-, Phenol, 4-[[4,6-bis(octylthio)-s-triazin-2-yl]amino]-2,6-di-tert-butyl- (7CI,8CI), 4-[[4,6-Bis(octylthio)-1,3,5-triazin-2-yl]amino]-2,6-bis(1,1-dimethylethyl)phenol, 1-(3,5-Di-tert-butyl-4-hydroxyanilino)-3,5-bis(octylthio)-s-triazine, 2,4-Bis(n-octylthio)-6-(3,5-di-tert-butyl-4-hydroxyphenylamino)-s-triazine, 2,4-Bis(n-octylthio)-6-(4'-hydroxy-3',5'-di-tert-butylanilino)-1,3,5-triazine, 2,4-Bis(octylthio)-6-(3,5-di-tert-butyl-4-hydroxyanilino)-s-triazine, 2,4-Bis(octylthio)-6-(4-hydroxy-3,5-di-tert-butylanilino)-1,3,5-triazine, 2,4-Bis(octylthio)-6-(4-hydroxy-3,5-di-tert-butylanilino)-s-triazine, 2,4-Bis(octylthio)-6-(4'-hydroxy-3',5'-di-tert-butylanilino)-1,3,5-triazine, 2,4-Bis-(n-octylthio)-6-(4-hydroxy-3,5-di-tert-butylanilino)-1,3,5-triazine, 2,4-Bis-(n-octylthio)-6-(4-hydroxy-3',5'-di-tert-butylanilino)-1,3,5-triazine, 2,6-Di-tert-butyl-4-[4,6-bis(octylthio)-1,3,5-triazin-2-ylamino]phenol, 2-(4-Hydroxy-3,5-di-tert-butylanilino)-4,6-bis(octylthio)-1,3,5-triazine, 2-(4-Hydroxy-3,5-di-tert-butylanilino)-4,6-bis(octylthio)-s-triazine, 2-(4-Hydroxy-3,5-di-tert-butylanilino)-4,6-di(octylthio)-s-triazine, 6-(3,5-Di-tert-butyl-4-hydroxyanilino)-2,4-bis(octylthio)-s-triazine, 6-(3,5-di-tert-Butyl-4-hydroxyanilino)-2,4-bis(n-octylthio)-s-triazine, 6-(4-Hydroxy-3,5-di-tert-butylanilino)-2,4-bis(n-octylthio)-1,3,5-triazine, 6-(4-Hydroxy-3,5-di-tert-butylanilino)-2,4-bis(octylthio)-1,3,5-triazine, 6-(4-Hydroxy-3,5-di-tert-butylanilino)-2,4-bis(octylthio)-s-triazine, Antioxidant 565, BNX 565, BOTAP, Eunox AO 565, Irganox 565, Irganox RA 565, NSC 328455, Tinogard MD 1
IUPAC Name 4-[[4,6-bis(octylsulfanyl)-1,3,5-triazin-2-yl]amino]-2,6-ditert-butylphenol
SMILES CCCCCCCCSc1nc(Nc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)nc(SCCCCCCCC)n1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C33H56N4OS2/c1-9-11-13-15-17-19-21-39-30-35-29(36-31(37-30)40-22-20-18-16-14-12-10-2)34-25-23-26(32(3,4)5)28(38)27(24-25)33(6,7)8/h23-24,38H,9-22H2,1-8H3,(H,34,35,36,37)
Formula Weight 588.3896
Chemical Name or Material 2,4-Bis(n-octylthio)-6-(4'-hydroxy-3',5'-di-tert-butylanilino)-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.