missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-[3-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane, ≥97%, Thermo Scientific™

Product Code. 10002214
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10002214 1 g Amber Glass Bottle 1g
10728835 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10002214 Supplier Thermo Scientific Maybridge Supplier No. CC63639DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 912569-68-7
Molecular Formula C19H23BO3
Molecular Weight (g/mol) 310.20
MDL Number MFCD09065022
InChI Key AFDZGEOQKHIWKF-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-3-phenoxymethyl phenyl-1,3,2-dioxaborolane
PubChem CID 24229702
IUPAC Name 4,4,5,5-tetramethyl-2-[3-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC(COC2=CC=CC=C2)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 310.2
Percent Purity 97%
Chemical Name or Material 4,4,5,5-Tetramethyl-2-[3-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.