missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-[2-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane, 97%, Thermo Scientific™

Catalog Number 10624595
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
912569-55-2
C19H23BO3
310.20
MFCD09064986
DTPMFYHICLWUGJ-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-2-phenoxymethyl phenyl-1,3,2-dioxaborolane
24229585
4,4,5,5-tetramethyl-2-[2-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CC=CC=C1COC1=CC=CC=C1
This item is not returnable. View return policy
912569-55-2
C19H23BO3
310.20
MFCD09064986
DTPMFYHICLWUGJ-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-2-phenoxymethyl phenyl-1,3,2-dioxaborolane
24229585
4,4,5,5-tetramethyl-2-[2-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CC=CC=C1COC1=CC=CC=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 310.2
Percent Purity 97%
Chemical Name or Material 4,4,5,5-Tetramethyl-2-[2-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'