missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4,5,5,5-Pentafluoropentan-1-ol, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Color | Colorless |
Boiling Point | 134°C to 135°C |
Flash Point | 63°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (GC) |
Quantity | 25 g |
Formula Weight | 178.1 |
Percent Purity | 95% |
Show More |
Safety and Handling
Product Identifier
- 4,4,5,5,5-Pentafluoropentan-1-ol
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P330-Rinse mouth.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'