missing translation for 'onlineSavingsMsg'
Learn More

4,4',4″-Trimethyltrityl alcohol, 98+%

Product Code. 11397685
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397685 25 g 25g
11387685 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397685 Supplier Thermo Scientific Alfa Aesar Supplier No. L02392.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3247-00-5 | C22H22O | 302.417 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3247-00-5
Molecular Formula C22H22O
Molecular Weight (g/mol) 302.417
MDL Number MFCD00014919
InChI Key DNWQXZDDISHGRM-UHFFFAOYSA-N
Synonym 4,4',4-trimethyltrityl alcohol, tris 4-methylphenyl methanol, methanol, tri-p-tolyl, tri-p-tolylmethanol, tri p-tolyl methanol, 4,4'-dimethyl-4-methyltrityl alcohol, benzenemethanol, 4-methyl-.alpha.,.alpha.-bis 4-methylphenyl, acmc-20amt7, tris 4-methylphenyl methan-1-ol, 4,4-trimethyltriphenylmethanol
PubChem CID 76733
IUPAC Name tris(4-methylphenyl)methanol
SMILES CC1=CC=C(C=C1)C(C2=CC=C(C=C2)C)(C3=CC=C(C=C3)C)O

Specifications

Melting Point 92°C to 96°C
Quantity 25 g
Beilstein 2055803
Formula Weight 302.42
Percent Purity ≥98%
Chemical Name or Material 4,4',4''-Trimethyltrityl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.