missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluorobutyric acid, 97%

Product Code. 11489727
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489727 5 g 5g
11479727 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489727 Supplier Thermo Scientific Alfa Aesar Supplier No. B22186.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 406-93-9 | C4H5F3O2 | 142.077 g/mol

4,4,4-Trifluorobutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4,4-Trifluorobutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 406-93-9
Molecular Formula C4H5F3O2
Molecular Weight (g/mol) 142.077
MDL Number MFCD00077604
InChI Key WTUCTMYLCMVYEX-UHFFFAOYSA-N
Synonym 4,4,4-trifluorobutyric acid, 4,4,4-trifluoro-butyric acid, butanoic acid, 4,4,4-trifluoro, zlchem 266, acmc-1ai6e, 4,4,4-trifluorobutyricacid, trifluoropropylcarboxylic acid, 4,4,4 trifluorobutyric acid, ksc237i6h, 3-trifluoromethylpropionic acid
PubChem CID 2777085
IUPAC Name 4,4,4-trifluorobutanoic acid
SMILES C(CC(F)(F)F)C(=O)O

Specifications

Melting Point 29°C to 32°C
Boiling Point 166°C to 167°C
Quantity 5 g
UN Number UN3261
Solubility Information Soluble in water.
Formula Weight 142.08
Percent Purity 97%
Chemical Name or Material 4,4,4-Trifluorobutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.