missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-3-hydroxy-3-methylbutyric acid, 98%

Product Code. 11383806
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383806 1 g 1g
11393806 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383806 Supplier Thermo Scientific Alfa Aesar Supplier No. L16873.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 338-03-4 | C5H7F3O3 | 172.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338-03-4
Molecular Formula C5H7F3O3
Molecular Weight (g/mol) 172.103
MDL Number MFCD00039531
InChI Key JCJGOWYCFRASTM-UHFFFAOYSA-N
Synonym 3-trifluoromethyl-3-hydroxybutyric acid, 4,4,4-trifluoro-3-hydroxy-3-methylbutyric acid, 3-hydroxy-3-methyl-4,4,4-trifluorobutyric acid, 4,4,4-trifluoro-3-hydroxy-3-methyl-butyric acid, butanoic acid, 4,4,4-trifluoro-3-hydroxy-3-methyl, pubchem12644, acmc-20an2r, 3-hydroxy-3-trifluoromethylbutyric acid, 3-hydroxy-3-trifluoromethyl butyric acid
PubChem CID 136168
IUPAC Name 4,4,4-trifluoro-3-hydroxy-3-methylbutanoic acid
SMILES CC(CC(=O)O)(C(F)(F)F)O

Specifications

Melting Point 39°C to 41°C
Boiling Point 87°C to 90°C (0.5 mmHg)
Refractive Index 1.388
Quantity 1 g
UN Number UN3261
Formula Weight 172.1
Percent Purity 98%
Chemical Name or Material 4,4,4-Trifluoro-3-hydroxy-3-methylbutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.