missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-1-phenyl-1,3-butanedione, 99%

Product Code. 10032800 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
Product Code. Quantity Packaging unitSize
10032800 25 g Glass bottle 25g
1 options
This item is not returnable. View return policy

Product Code. 10032800

Brand: Thermo Scientific Acros 202670250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 326-06-7 | C10H7F3O2 | 216.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 326-06-7
CAS Min % 98.5
CAS Max % 100.0
Melting Point 37.0°C to 40.0°C
Color White to Yellow
Boiling Point 224.0°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C10H7F3O2
Linear Formula C6H5COCH2COCF3
MDL Number MFCD00000425
Quantity 25 g
Beilstein 07, III, 3490
Synonym benzoyl-1,1,1-trifluoroacetone, benzoyltrifluoroacetone, 4,4,4-trifluoro-1-phenyl-1,3-butanedione, benzoyl trifluoroacetyl methane, 3-benzoyl-1,1,1-trifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-phenyl, 4,4,4-trifluoro-1-phenyl-butane-1,3-dione, 1-benzoyl-3,3,3-trifluoroacetone, .omega.-trifluoroacetyl acetophenone, 1-benzoyl-3,3,3-trifluoro-2-propanone
Solubility Information Solubility in water: practically insoluble. Other solubilities: practically insoluble in higher alcohol,easily soluble in lower alcohol
InChI Key VVXLFFIFNVKFBD-UHFFFAOYSA-N
SMILES FC(F)(F)C(=O)CC(=O)C1=CC=CC=C1
IUPAC Name 4,4,4-trifluoro-1-phenylbutane-1,3-dione
Molecular Weight (g/mol) 216.16
PubChem CID 67589
Formula Weight 216.16
Percent Purity 99%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4,4,4-Trifluoro-1-phenyl-1,3-butanedione, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.