missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione, TRC

Product Code. 30454291
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454291 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454291 Supplier Toronto Research Chemicals Supplier No. TRCT7871001G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione
CAS 582-65-0
Molecular Formula C10H6F4O2
Quantity 1 g
Synonym 1,3-Butanedione','4,4,4-trifluoro-1-(p-fluorophenyl)- (6CI','8CI), 4,4,4-Trifluoro-1-(4-fluorophenyl)-1,3-butanedione (ACI), 1,1,1-Trifluoro-4-(4-fluorophenyl)-2,4-butanedione, 1-(4-Fluorophenyl)-4,4,4-trifluoro-1,3-butanedione, 1-(4′-Fluorophenyl)-4,4,4-trifluoro-1,3-butanedione, 4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione
InChI Formula InChI=1S/C10H6F4O2/c11-7-3-1-6(2-4-7)8(15)5-9(16)10(12,13)14/h1-4H,5H2
SMILES O=C(C1=CC=C(F)C=C1)CC(C(F)(F)F)=O
IUPAC Name 4,4,4-trifluoro-1-(4-fluorophenyl)butane-1,3-dione
Molecular Weight (g/mol) 234.15
Formula Weight 234.03
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.