missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione, TRC

Product Code. 30453378
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453378 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453378 Supplier Toronto Research Chemicals Supplier No. TRCT7871002.5G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione
CAS 582-65-0
Molecular Formula C10H6F4O2
Quantity 2.5 g
Synonym 1,3-Butanedione','4,4,4-trifluoro-1-(p-fluorophenyl)- (6CI','8CI), 4,4,4-Trifluoro-1-(4-fluorophenyl)-1,3-butanedione (ACI), 1,1,1-Trifluoro-4-(4-fluorophenyl)-2,4-butanedione, 1-(4-Fluorophenyl)-4,4,4-trifluoro-1,3-butanedione, 1-(4′-Fluorophenyl)-4,4,4-trifluoro-1,3-butanedione, 4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione
InChI Formula InChI=1S/C10H6F4O2/c11-7-3-1-6(2-4-7)8(15)5-9(16)10(12,13)14/h1-4H,5H2
SMILES O=C(C1=CC=C(F)C=C1)CC(C(F)(F)F)=O
IUPAC Name 4,4,4-trifluoro-1-(4-fluorophenyl)butane-1,3-dione
Molecular Weight (g/mol) 234.15
Formula Weight 234.03
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.