missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4,4-Trifluoro-1-(2-furyl)-1,3-butanedione, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 20°C to 21°C |
| Density | 1.391 |
| Boiling Point | 203°C |
| Flash Point | 87°C (188°F) |
| Refractive Index | 1.528 |
| Quantity | 5 g |
| Beilstein | 168085 |
| Formula Weight | 206.12 |
| Percent Purity | 98% |
| Chemical Name or Material | 4,4,4-Trifluoro-1- (2-furyl)-1,3-butanedione |
Safety and Handling
Product Identifier
- 4,4,4-Trifluoro-1-(2-furyl)-1,3-butanedione
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: 4,4,4-Trifluoro-1-(2-furyl)-1,3-butanedione
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?