missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-1-(2-furyl)-1,3-butanedione, 98%

Product Code. 11404290
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404290 25 g 25g
11494280 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404290 Supplier Thermo Scientific Alfa Aesar Supplier No. A10693.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 326-90-9 | C8H5F3O3 | 206.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 326-90-9
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD00020935
InChI Key OWLPCALGCHDBCN-UHFFFAOYSA-N
Synonym 4,4,4-trifluoro-1-furan-2-yl butane-1,3-dione, 2-furoyltrifluoroacetone, furoyltrifluoroacetone, 4,4,4-trifluoro-1-2-furyl-1,3-butanedione, perfluoroacetyl 2-furoyl methane, furonyltrifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-furanyl, 4,4,4-trifluoro-1-2-furyl butane-1,3-dione, 2-furoyl trifluoro acetone, 4,4,4-trifluoro-1-2-furanyl-1,3-butanedione
PubChem CID 67594
IUPAC Name 4,4,4-trifluoro-1-(furan-2-yl)butane-1,3-dione
SMILES FC(F)(F)C(=O)CC(=O)C1=CC=CO1

Specifications

Melting Point 20°C to 21°C
Density 1.391
Boiling Point 203°C
Flash Point 87°C (188°F)
Refractive Index 1.528
Quantity 25 g
Beilstein 168085
Formula Weight 206.12
Percent Purity 98%
Chemical Name or Material 4,4,4-Trifluoro-1- (2-furyl)-1,3-butanedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.