missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)tetrahydropyran, Thermo Scientific™

Product Code. 10240220
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10240220 5 g 5g
10526070 250 mg 250mg
10109419 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10240220 Supplier Thermo Scientific Maybridge Supplier No. CC29939DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1131912-76-9
Molecular Formula C11H21BO3
Molecular Weight (g/mol) 212.10
MDL Number MFCD11506065
InChI Key NLSMOSUUBUCSPL-UHFFFAOYSA-N
Synonym tetrahydropyran-4-boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-tetrahydro-2h-pyran-4-yl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl tetrahydropyran, tetrahydropyran-4-boronic acid, pinacol ester, tetrahydropyran-4-boronicacidpinacolester, 4,4,5,5-tetramethyl-2-oxan-4-yl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl oxane, tetrahydro-2h-pyran-4-yl boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl tetrahydro-2h-pyran
PubChem CID 42614649
IUPAC Name 4,4,5,5-tetramethyl-2-(oxan-4-yl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1CCOCC1

Specifications

Assay Percent Range 97%
Quantity 5 g
Formula Weight 212.09
Percent Purity 97%
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)tetrahydropyran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.