missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 97%, Maybridge

Product Code. 10235804
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10235804 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10235804 Supplier Thermo Scientific Maybridge Supplier No. MAY00013DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 568577-88-8
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C16H24BNO3
MDL Number MFCD04112544
Quantity 1 g
Synonym 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 4-morpholinophenylboronic acid pinacol ester, 4-morpholinophenylboronic acid, pinacol ester, 4-morpholin-4-yl-phenylboronic acid pinacol ester, 4-morpholin-4-ylbenzeneboronic acid pinacol ester, morpholine, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl, 4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 4-morpholino phenylboronic acid pinacolate, 4-4-morpholinyl benzeneboronic acid pinacol ester
InChI Key UCPALIMHMYIZPZ-UHFFFAOYSA-N
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3CCOCC3
IUPAC Name 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
Molecular Weight (g/mol) 289.182
PubChem CID 2795361
Formula Weight 289.18
Percent Purity 97%
Chemical Name or Material 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.