missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(1-Hydroxyethyl)-2-methoxy-5-nitrophenoxy]butanoic Acid, TRC

Product Code. 17913285
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17913285 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17913285 Supplier Toronto Research Chemicals Supplier No. TRCH8252551G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 175281-76-2
Molecular Formula C13 H17 N O7
Quantity 1 g
InChI Formula InChI=1S/C13H17NO7/c1-8(15)9-6-11(20-2)12(7-10(9)14(18)19)21-5-3-4-13(16)17/h6-8,15H,3-5H2,1-2H3,(H,16,17)
SMILES COc1cc(C(C)O)c(cc1OCCCC(=O)O)[N+](=O)[O-]
IUPAC Name 4-[4-(1-hydroxyethyl)-2-methoxy-5-nitrophenoxy]butanoic acid
Molecular Weight (g/mol) 299.2766
Formula Weight 299.1005
Chemical Name or Material 4-[4-(1-Hydroxyethyl)-2-methoxy-5-nitrophenoxy]butanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.