missing translation for 'onlineSavingsMsg'
Learn More

4(3H)-Quinazolinone, MedChemExpress

Product Code. 16409406
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16409406

Brand: Medchem Express HYW018800

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 4(3H)-Quinazolinone is a building block in chemical synthesis. Biologically active nitrogen heterocyclic compounds. Possesses a wide spectrum of biological properties like antibacterial, antifungal, anticonvulsant, anti-inflammatory, anti-HIV, anticancerous and analgesic activities.

4(3H)-Quinazolinone is a building block in chemical synthesis. Biologically active nitrogen heterocyclic compounds. Possesses a wide spectrum of biological properties like antibacterial, antifungal, anticonvulsant, anti-inflammatory, anti-HIV, anticancerous and analgesic activities.

Specifications

Chemical Name or Material 4(3H)-Quinazolinone
Physical Form Solid
CAS 491-36-1
Color White to Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H6N2O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 110 mg/mL (752.65 mM; Need ultrasonic) ∣H2O : 1.2 mg/mL (8.21 mM; Need ultrasonic)
SMILES O=C1NC=NC2=C1C=CC=C2
Molecular Weight (g/mol) 146.15
Formula Weight 146.15
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.