missing translation for 'onlineSavingsMsg'
Learn More

4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10753512
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10753512 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10753512 Supplier Thermo Scientific Maybridge Supplier No. MAY00318DA
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 874839-13-1
Assay Percent Range 97%
Molecular Formula C12H7ClF3NO3S
Quantity 1 g
Synonym 4-3-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 4-3-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, chloro 4-3-trifluoromethyl 2-pyridyloxy phenyl sulfone, 4-3-trifluoromethyl pyridin-2-yl oxy benzenesulphonyl chloride
InChI Key FDAYBQVWHDJJOL-UHFFFAOYSA-N
SMILES C1=CC(=C(N=C1)OC2=CC=C(C=C2)S(=O)(=O)Cl)C(F)(F)F
IUPAC Name 4-[3-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride
Molecular Weight (g/mol) 337.697
PubChem CID 45594320
Formula Weight 337.7
Chemical Name or Material 4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.