missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Trifluoromethyl)phenyl]-4-piperidinol, 97%, Thermo Scientific™

Product Code. 10225380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10225380 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10225380 Supplier Thermo Scientific Acros Supplier No. 136980250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Specifications

CAS 2249-28-7
CAS Min % 96.0
CAS Max % 100.0
Melting Point 91°C to 95°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C12H14F3NO
MDL Number MFCD00044832
Quantity 25 g
Synonym 4-3-trifluoromethyl phenyl-4-piperidinol, 4-3-trifluoromethyl phenyl piperidin-4-ol, 4-piperidinol, 4-3-trifluoromethyl phenyl, 4-alpha,alpha,alpha-trifluoro-m-tolyl piperidin-4-ol, 4-3-trifuoromethyl phenyl-4-piperidinol, 4-3-trifluoromethylphenyl-4-piperidinol, 4-pound 3-trifluoromethylphenyl-4-piperidinol, 4-3-trifluoromethylphenyl-4'-piperidinol, 4-3-trifluoromethyl phenyl-4'-piperidinol
InChI Key ILKPZCKFWLTEBQ-UHFFFAOYSA-N
SMILES C1CNCCC1(C2=CC(=CC=C2)C(F)(F)F)O
IUPAC Name 4-[3-(trifluoromethyl)phenyl]piperidin-4-ol
Molecular Weight (g/mol) 245.245
PubChem CID 75255
Formula Weight 245.24
Percent Purity ≥96.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-[3-(Trifluoromethyl)phenyl]-4-piperidinol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4- 3-(Trifluoromethyl)phenyl]-4-piperidinol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.