missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride, 97%

Product Code. 15498485
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15498485 5 g 5g
15408495 250 mg 250mg
15488485 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15498485 Supplier Thermo Scientific Alfa Aesar Supplier No. H33473.06
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 874839-13-1 | C12H7ClF3NO3S | 337.697 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874839-13-1
Molecular Formula C12H7ClF3NO3S
Molecular Weight (g/mol) 337.697
MDL Number MFCD07774164
InChI Key FDAYBQVWHDJJOL-UHFFFAOYSA-N
Synonym 4-3-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 4-3-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, chloro 4-3-trifluoromethyl 2-pyridyloxy phenyl sulfone, 4-3-trifluoromethyl pyridin-2-yl oxy benzenesulphonyl chloride
PubChem CID 45594320
IUPAC Name 4-[3-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride
SMILES C1=CC(=C(N=C1)OC2=CC=C(C=C2)S(=O)(=O)Cl)C(F)(F)F

Specifications

Melting Point 63°C to 65°C
Quantity 5 g
UN Number UN3261
Formula Weight 337.71
Percent Purity 97%
Chemical Name or Material 4-(3-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.