missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Thienyl)benzaldehyde, ≥97%, Thermo Scientific™

Product Code. 10235524
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10235524 1 g 1g
10652315 250 mg 250mg
10527104 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10235524 Supplier Thermo Scientific Maybridge Supplier No. CC48704DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 157730-74-0
Molecular Formula C11H8OS
Molecular Weight (g/mol) 188.244
MDL Number MFCD04039151
InChI Key MGPZYZLJJSEFLU-UHFFFAOYSA-N
PubChem CID 3280631
IUPAC Name 4-thiophen-3-ylbenzaldehyde
SMILES C1=CC(=CC=C1C=O)C2=CSC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 4-thiophen-3-yl benzaldehyde, 4-3-thienyl benzaldehyde, 4-3-thienyl benzenecarbaldehyde, 4-thiophen-3-yl-benzaldehyde, benzaldehyde,4-3-thienyl, 4-thien-3-yl benzaldehyde, pubchem15939, thienylbenzenecarbaldehyde, 4-3-thiophenyl benzaldehyde, benzaldehyde, 4-3-thienyl
Formula Weight 188.25
Percent Purity 97%
Chemical Name or Material 4-(3-Thienyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.