missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Pyrrolidinyl)morpholine, 97%

Product Code. 11429790
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429790 1 g 1g
11419790 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429790 Supplier Thermo Scientific Alfa Aesar Supplier No. H51002.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 53617-37-1 | C8H16N2O | 156.23 g/mol

4-(3-Pyrrolidinyl)morpholine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(3-Pyrrolidinyl)morpholine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53617-37-1
Molecular Formula C8H16N2O
Molecular Weight (g/mol) 156.23
MDL Number MFCD06740352
InChI Key OPJDRNMJJNFSNZ-UHFFFAOYNA-N
Synonym 4-pyrrolidin-3-yl morpholine, 4-pyrrolidin-3-yl-morpholine, 4-3-pyrrolidinyl morpholine, 4-3-pyrrolidino morpholine, acmc-20aiao, pubchem11379, 3-morpholin-4-yl pyrrolidine, morpholine, 4-3-pyrrolidinyl
PubChem CID 14627373
IUPAC Name 4-pyrrolidin-3-ylmorpholine
SMILES C1CC(CN1)N1CCOCC1

Specifications

Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 156.23
Percent Purity 97%
Chemical Name or Material 4-(3-Pyrrolidinyl)morpholine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.