missing translation for 'onlineSavingsMsg'
Learn More

4-[(3-Oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid, Alfa Aesar™

Product Code. 11449350
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449350 1 g 1g
11439350 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449350 Supplier Thermo Scientific Alfa Aesar Supplier No. H50234.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 857492-98-9
Molecular Formula C16H13NO4
Molecular Weight (g/mol) 283.283
MDL Number MFCD00269588
InChI Key YVNJRPNQFNUCKP-UHFFFAOYSA-N
Synonym 4-3-oxo-2,3-dihydro-4h-1,4-benzoxazin-4-yl methyl benzoic acid, 4-2,3-dihydro-3-oxo-4h-1,4-benzoxazin-4-yl methyl benzoic acid, 4-4-carboxybenzyl-2h-1,4-benzoxazin-3 4h-one, 4-3-oxo-2h-1,4-benzoxazin-4-yl methyl benzoic acid, 4-3-oxo-2h-benzo e 1,4-oxazin-4-yl methyl benzoic acid, acmc-20amr3, 4-3-oxo-1,4-benzoxazin-4-yl methyl benzoic acid, 4-3-oxo-2h-benzo b 1,4 oxazin-4 3h-yl methyl benzoic acid, benzoic acid,4-2,3-dihydro-3-oxo-4h-1,4-benzoxazin-4-yl methyl
PubChem CID 1659722
IUPAC Name 4-[(3-oxo-1,4-benzoxazin-4-yl)methyl]benzoic acid
SMILES C1C(=O)N(C2=CC=CC=C2O1)CC3=CC=C(C=C3)C(=O)O

Specifications

Quantity 1 g
Formula Weight 283.29
Chemical Name or Material 4-[(3-Oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.