missing translation for 'onlineSavingsMsg'
Learn More

4-[(3-Isopropyl-2-oxo-1,3-oxazolidin-5-yl)methoxy]benzaldehyde (Bisoprolol Oxazolidinone-benzaldehyde), Mikromol™

Product Code. 30052061
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052061 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30052061 Supplier Mikromol Supplier No. MM0460.170025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-[(3-Isopropyl-2-oxo-1,3-oxazolidin-5-yl)methoxy]benzaldehyde (Bisoprolol Oxazolidinone-benzaldehyde)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[(3-Isopropyl-2-oxo-1,3-oxazolidin-5-yl)methoxy]benzaldehyde (Bisoprolol Oxazolidinone-benzaldehyde)
CAS 1173203-26-3
Analyte or Component Names 4-[(3-Isopropyl-2-oxo-1,3-oxazolidin-5-yl)methoxy]benzaldehyde (Bisoprolol Oxazolidinone-benzaldehyde)
Shipping Condition Room Temperature
Molecular Formula C14 H17 N O4
Certifications/Compliance ISO 17025
Synonym 4-[(3-Isopropyl-2-oxo-1,3-oxazolidin-5-yl)methoxy]benzaldehyde, Bisoprolol Fumarate Imp. T (EP), Bisoprolol Oxazolidinone-benzaldehyde, Bisoprolol Imp. T (EP), Bisoprolol Fumarate Impurity T, Bisoprolol Impurity T
InChI Formula InChI=1S/C14H17NO4/c1-10(2)15-7-13(19-14(15)17)9-18-12-5-3-11(8-16)4-6-12/h3-6,8,10,13H,7,9H2,1-2H3
SMILES CC(C)N1CC(COc2ccc(C=O)cc2)OC1=O
IUPAC Name 4-[(2-oxo-3-propan-2-yl-1,3-oxazolidin-5-yl)methoxy]benzaldehyde
Molecular Weight (g/mol) 263.29
Quantity 25 mg
Formula Weight 263.1158 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.