missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Isopropyl-[1,2,4]oxadiazol-5-yl)piperidine-1-carboxylic acid tert-butyl ester, Thermo Scientific™

Product Code. 10687580
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10687580 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10687580 Supplier Thermo Scientific Maybridge Supplier No. MAY00254DA
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 913264-42-3
Packaging Amber Glass Bottle
Molecular Formula C15H25N3O3
MDL Number MFCD11226634
Quantity 1 g
Synonym tert-butyl 4-3-isopropyl-1,2,4-oxadiazol-5-yl piperidine-1-carboxylate, 4-3-isopropyl-1,2,4 oxadiazol-5-yl piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-3-isopropyl-1,2,4-oxadiazol-5-yl piperidin-1-carboxylate, tert-butyl4-3-isopropyl-1,2,4-oxadiazol-5-yl piperidine-1-carboxylate, tert-butyl 4-3-propan-2-yl-1,2,4-oxadiazol-5-yl piperidine-1-carboxylate
InChI Key QIHZUQZZJHBESB-UHFFFAOYSA-N
SMILES CC(C)C1=NOC(=N1)C1CCN(CC1)C(=O)OC(C)(C)C
IUPAC Name tert-butyl 4-(3-propan-2-yl-1,2,4-oxadiazol-5-yl)piperidine-1-carboxylate
Molecular Weight (g/mol) 295.38
PubChem CID 37859644
Formula Weight 295.38
Percent Purity 95%
Chemical Name or Material 4-(3-Isopropyl-[1, 2, 4]oxadiazol-5-yl)piperidine-1-carboxylic acid tert-butyl ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.