missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Hydroxypropyl)phenyl beta-D-Glucopyranosiduronic Acid, TRC

Product Code. 30402587
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402587 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402587 Supplier Toronto Research Chemicals Supplier No. TRCH95279550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(3-Hydroxypropyl)phenyl beta-D-Glucopyranosiduronic Acid
CAS 549548-40-5
Molecular Formula C15H20O8
Quantity 50 mg
InChI Formula InChI=1S/C15H20O8/c16-7-1-2-8-3-5-9(6-4-8)22-15-12(19)10(17)11(18)13(23-15)14(20)21/h3-6,10-13,15-19H,1-2,7H2,(H,20,21)/t10-,11-,12+,13-,15+/m0/s1
SMILES OCCCc1ccc(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)cc1
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[4-(3-hydroxypropyl)phenoxy]oxane-2-carboxylic acid
Molecular Weight (g/mol) 328.31
Formula Weight 328.1158
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.